missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL5/RANTES Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 16 publications
Brand: R&D Systems MAB478-500
This item is not returnable.
View return policy
Description
CCL5/RANTES Monoclonal specifically detects CCL5/RANTES in Mouse samples. It is validated for Neutralization, Immunocytochemistry.
Specifications
| CCL5/RANTES | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6352 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization, Immunocytochemistry | |
| 53405 | |
| Neutralization 0.1-0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P30882 | |
| CCL5 | |
| E. coli-derived recombinant mouse CCL5/RANTES Ser24-Ser91 Accession # P30882 | |
| 500 μg | |
| Primary | |
| Detects mouse CCL5/RANTES in direct ELISAs. Shows 50% cross-reactivity with recombinant human (rh) CCL5, and no cross-reactivity with recombinant feline CCL5, rhCCL3, recombinant mouse (rm) CCL3, rmCCL4, rhCCL4, rhCCL14, or rhCCL18. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction