missing translation for 'onlineSavingsMsg'
Learn More

Mouse CCL5/RANTES Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18732688
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732688 25 μg 25µg
18718399 100 μg 100µg
18783017 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18732688 Supplier R&D Systems Supplier No. MAB478SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 16 publications

CCL5/RANTES Monoclonal specifically detects CCL5/RANTES in Mouse samples. It is validated for Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL5/RANTES
Applications Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 53405
Conjugate Unconjugated
Dilution Neutralization 0.1-0.5 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P30882
Gene Alias chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Gene Symbols CCL5
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL5/RANTES Ser24-Ser91 Accession # P30882
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6352
Test Specificity Detects mouse CCL5/RANTES in direct ELISAs. Shows 50% cross-reactivity with recombinant human (rh) CCL5, and no cross-reactivity with recombinant feline CCL5, rhCCL3, recombinant mouse (rm) CCL3, rmCCL4, rhCCL4, rhCCL14, or rhCCL18.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.