missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CCL4/MIP-1 beta Quantikine ELISA Kit
Description
The Mouse CCL4/MIP-1 beta Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL4/MIP-1 beta in cell culture supernates (50 ul), serum (25 ul), edta plasma (25 ul).
Specifications
Specifications
| Assay Range | 7.80 - 500pg/mL |
| Assay Sensitivity | 3pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Serum |
| Specificity | Natural and Recombinant Mouse MIP-1 beta |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20303 |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (50uL),Serum (25uL),EDTA Plasma (25uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?