missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL4/MIP-1 beta Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB451-100
This item is not returnable.
View return policy
Description
CCL4/MIP-1 beta Monoclonal specifically detects CCL4/MIP-1 beta in Mouse samples. It is validated for Neutralization.
Specifications
| CCL4/MIP-1 beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6351 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization | |
| 46907 | |
| Neutralization 1-3 ug/mL | |
| Q5QNV9 | |
| CCL4 | |
| E. coli-derived recombinant mouse CCL4/MIP-1 beta Ala24-Asn92 Accession # Q5QNV9 | |
| 100 μg | |
| Primary | |
| Detects mouse CCL4/MIP-1 beta in ELISAs. Does not cross-react with recombinant human (rh) CCL1, 2, 4/MIP-1 alpha , 4/MIP-1 beta , 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, rmCCL1, 2, 4/MIP-1 alpha , 4/MIP-1 beta , 5, 6, 7, 9/10, 11, 12, 17, 19, 20, 21, 22, 24, 25, or rrCCL20. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction