missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL22/MDC Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£167.00 - £1535.00
Specifications
| Antigen | CCL22/MDC |
|---|---|
| Clone | 158132 |
| Dilution | Neutralization 0.4-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720658
![]() |
R&D Systems
MAB4391-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18775788
![]() |
R&D Systems
MAB4391-100 |
100 μg |
£389.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742477
![]() |
R&D Systems
MAB4391-500 |
500 μg |
£1535.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CCL22/MDC | |
| Neutralization 0.4-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| O88430 | |
| 6367 | |
| E. coli-derived recombinant mouse CCL22/MDC Gly25-Ser92 Accession # O88430 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL22/MDC in ELISAs and Western blots. In Western blots, this antibody shows 25% cross-reactivity with recombinant viral MIP-II and no cross-reactivity with rmCCL1, 2, 3, 4, 6, 7, CCL9/10/MIP-1 gamma , 11, 12, 19, 20, 21, 22, 24, 25, 27, rhCCL1, 2, 3, 4, 5, 7, 8, 11, 13, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 28, or rrCCL20. |
| 158132 | |
| Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
| CCL22 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title