missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL22/MDC Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF439-SP
This item is not returnable.
View return policy
Description
CCL22/MDC Polyclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL22/MDC | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6367 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.3-1.0 ug/mL | |
| Q9QZU2 | |
| CCL22 | |
| E. coli-derived recombinant mouse CCL22/MDC Gly25-Ser92 Accession # Q9QZU2 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL22/MDC in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human CCL22/MDC is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion