missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit

Product Code. 18772114
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
Unit Size:
1 set
Pack of 96
This item is not returnable. View return policy

Product Code. 18772114

Brand: R&D Systems™ SMJE00B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit quantifies mouse CCL2/JE/MCP-1 in cell culture supernates (50 ul), tissue lysates (50 ul), tissue culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

1 Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.81 - 500 pg/mL
Assay Sensitivity 0.666 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates
Specificity Natural and Recombinant Mouse MCP-1
For Use With (Application) ELISA
Gene ID (Entrez) 6347
Gene Symbol CCL2
Host Species Mouse
Quantity 1 Pack (6 Plates)
Sample Volume Cell Culture Supernates (50 uL),Tissue Lysates (50 uL),Tissue Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target CCL2/JE/MCP-1
Total Assay Time 4.5 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.