missing translation for 'onlineSavingsMsg'
Learn More

Mouse CCL2/JE/MCP-1 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 16 publications

£180.00 - £1775.00

Specifications

Antigen CCL2/JE/MCP-1
Clone 123616
Dilution Neutralization 0.75-3.0 ug/mL
Applications Neutralization
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18748709
GREENER_CHOICE
View Documents
R&D Systems
MAB479-SP
25 μg
£180.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18715129
GREENER_CHOICE
View Documents
R&D Systems
MAB479-100
100 μg
£448.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18713789
GREENER_CHOICE
View Documents
R&D Systems
MAB479-500
500 μg
£1775.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Neutralization.
Specifications

Specifications

CCL2/JE/MCP-1
Neutralization 0.75-3.0 ug/mL
Monoclonal
Rat
Mouse
P10148
6347
E. coli-derived recombinant mouse CCL2/JE/MCP-1 Gln24-Arg96 Accession # P10148
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse CCL2/JE/MCP-1 in ELISAs. Does not cross-react with recombinant human (rh) CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, rmCCL1, 3, 4, 5, 6, 7, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, 28, or rrCCL20.
123616
Neutralization
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Ccl2
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.