missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL19/MIP-3 beta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF880
This item is not returnable.
View return policy
Description
CCL19/MIP-3 beta Polyclonal specifically detects CCL19/MIP-3 beta in Mouse samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| CCL19/MIP-3 beta | |
| Polyclonal | |
| Unconjugated | |
| Q548P0 | |
| Ccl19 | |
| E. coli-derived recombinant mouse CCL19/MIP-3 beta Gly26-Val107 (Ser108LeuGlu) Accession # Q548P0 | |
| 100 μg | |
| Primary | |
| Detects mouse CCL19/MIP-3 beta in direct ELISAs and Western blots. In direct ELISAs, less than 30% cross-reactivity with recombinant rat CCL19/MIP-3 beta is observed and less than 10% cross-reactivity with recombinant human CCL19/MIP-3 beta is observed. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.8-4 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELC, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP3 beta, MIP-3 beta, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6363 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction