missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL17/TARC Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB5291
This item is not returnable.
View return policy
Description
CCL17/TARC Monoclonal specifically detects CCL17/TARC in Mouse samples. It is validated for Western Blot.
Specifications
| CCL17/TARC | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6361 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 110946 | |
| Western Blot 1 ug/mL | |
| Q9WUZ6 | |
| CCL17 | |
| E. coli-derived recombinant mouse CCL17/TARC Ala24-Pro93 Accession # Q9WUZ6 | |
| 500 μg | |
| Primary | |
| Detects mouse CCL17/TARC in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) CCL8 is observed and no cross-reactivity with rhCCL1, 2, 3, 4, 5, 7, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 28, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 9, 11, 12, 19, 20, 21, 22, 24, 25, 27, 28, or recombinant rat CCL20 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction