missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL12/MCP-5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4281-SP
This item is not returnable.
View return policy
Description
CCL12/MCP-5 Monoclonal specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Neutralization.
Specifications
| CCL12/MCP-5 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.5-2.5 ug/mL | |
| Q62401 | |
| Ccl12 | |
| E. coli-derived recombinant mouse CCL12/MCP-5 Gly23-Gly104 Accession # Q62401 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL12/MCP-5 in direct ELISAs. | |
| Mouse | |
| Purified |
| Neutralization | |
| 147801 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP5, MCP-5, Scya12 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 20293 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction