missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL12/MCP-5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF428-SP
This item is not returnable.
View return policy
Description
CCL12/MCP-5 Polyclonal specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL12/MCP-5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP5, MCP-5, Scya12 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 20293 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.8-4.0 ug/mL | |
| Q62401 | |
| Ccl12 | |
| E. coli-derived recombinant mouse CCL12/MCP-5 (R&D Systems, Catalog # 428-P5) Gly23-Gly104 Accession # Q62401 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL12/MCP-5 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) MCP-3 is observed and 5% cross-reactivity with recombinant mouse (rm) JE, rmMARC, rhMCP-1, and rhMCP-2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction