missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL11/Eotaxin Biotinylated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems BAM420
This item is not returnable.
View return policy
Description
CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CCL11/Eotaxin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P48298 | |
| CCL11 | |
| E. coli-derived recombinant mouse CCL11/Eotaxin His24-Pro97 Accession # P48298 | |
| 250 μg | |
| Primary | |
| Detects mouse CCL11/Eotaxin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human (rh) Eotaxin, recombinant mouse MARC, and rh6Ckine is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 42285 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6356 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto