missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL1/I-309/TCA-3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF845
This item is not returnable.
View return policy
Description
Produced in goats immunized with purified, Sf 21-derived, recombinant mouse CCL1 (rmCCL1). CCL1 specific IgG was purified by mouse CCL1 affinity chromatography.
CCL1/I-309/TCA-3 Polyclonal specifically detects CCL1/I-309/TCA-3 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL1/I-309/TCA-3 | |
| Polyclonal | |
| Unconjugated | |
| P10146 | |
| CCL1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CCL1/I-309/TCA-3 Lys24-Cys92 Accession # P10146 | |
| 100 μg | |
| Primary | |
| Detects mouse CCL1 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.3-1.5 ug/mL | |
| C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1, SCYA1Small-inducible cytokine A1, SCYA2, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3, TCA-3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6346 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction