missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin E Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1130-SP
This item is not returnable.
View return policy
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Mouse samples. It is validated for Western Blot.
Specifications
| Cathepsin E | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P70269 | |
| CTSE | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin E Gln19-Pro397 Accession # P70269 | |
| 25 μg | |
| Primary | |
| Detects mouse Cathepsin E in direct ELISAs and Western blots. It recognizes both the pro and active forms of recombinant mouse (rm) Cathepsin E. In Western blots, no cross-reactivity with rmCathepsin B, C, H, L, or recombinant human Cathepsin O, S, or V is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 186513 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1510 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction