missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mouse Caspase-10/FLICE2 ELISA Kit (Chemiluminescence)
Shop All Bio Techne ProductsDescription
The Mouse Caspase-10/FLICE2 ELISA Kit (Chemiluminescence) measures mouse Caspase-10/FLICE2 in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 7.81-500 pg/mL |
| Assay Sensitivity | 4.69 pg/mL |
| Cross Reactivity | This kit recognizes Mouse CASP10 in samples. No significant cross-reactivity or interference between Mouse CASP10 and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 843 |
| Gene Symbol | CASP10 |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?