missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Carboxypeptidase B1/CPB1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2898
This item is not returnable.
View return policy
Description
Carboxypeptidase B1/CPB1 Polyclonal specifically detects Carboxypeptidase B1/CPB1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Carboxypeptidase B1/CPB1 | |
| Polyclonal | |
| Unconjugated | |
| NP_083982 | |
| CPB1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase B1/CPB1 Ser16-Tyr415 Accession # NP_083982 | |
| 100 μg | |
| Primary | |
| Detects mouse Carboxypeptidase B1/CPB1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) CPB1 is observed and less than 1% cross-reactivity with rhCPA1 and recombinant mouse CPA4 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, CPB1, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1360 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction