missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Carboxylesterase 1/CES1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7929-SP
This item is not returnable.
View return policy
Description
Carboxylesterase 1/CES1 Polyclonal specifically detects Carboxylesterase 1/CES1 in Mouse samples. It is validated for Western Blot.
Specifications
| Carboxylesterase 1/CES1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1, SES1TGH, TGH, Triacylglycerol hydrolase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1066 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8VCC2 | |
| Ces1g | |
| Mouse myeloma cell line NS0-derived recombinant mouse Carboxylesterase 1/CES1 His19-Leu565 Accession # Q8VCC2 | |
| 25 μg | |
| Primary | |
| Detects mouse Carboxylesterase 1/CES1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse CES3 and recombinant human CES1 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction