missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Carbonic Anhydrase XII/CA12 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£167.00 - £442.00
Specifications
| Antigen | Carbonic Anhydrase XII/CA12 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704757
![]() |
R&D Systems
AF2345-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18082432
![]() |
R&D Systems
AF2345 |
100 μg |
£442.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase XII/CA12 Polyclonal specifically detects Carbonic Anhydrase XII/CA12 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Carbonic Anhydrase XII/CA12 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| CA12, Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
| Car12 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q8CI85 | |
| 771 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase XII/CA12 Ala25-Ser301 Accession # Q8CI85 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Carbonic Anhydrase XII/CA12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human CA12 is observed and less than 5% cross-reactivity with recombinant human CA1, rhCA2, rhCA3, rhCA8, recombinant mouse (rm) CA4, and rmCA9 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title