missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Carbonic Anhydrase XII/CA12 Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £346.00
Specifications
| Antigen | Carbonic Anhydrase XII/CA12 |
|---|---|
| Clone | 336103 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758069
![]() |
R&D Systems
MAB2345-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18160581
|
R&D Systems
MAB2345 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase XII/CA12 Monoclonal specifically detects Carbonic Anhydrase XII/CA12 in Mouse samples. It is validated for Western Blot.Specifications
| Carbonic Anhydrase XII/CA12 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8CI85 | |
| 771 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase XII/CA12 Ala25-Ser301 Accession # Q8CI85 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Carbonic Anhydrase XII/CA12 in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human (rh) CA12, 10% cross-reactivity with rhCA2, and no cross-reactivity with rhCA1, 3, 5A, 5B, 6, 7, 8, 10, 11, 13, recombinant mouse CA4, 9, or 14 is observed. |
| 336103 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CA12, Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
| Car12 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title