missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cadherin-4/R-Cadherin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6677
This item is not returnable.
View return policy
Description
Cadherin-4/R-Cadherin Polyclonal specifically detects Cadherin-4/R-Cadherin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cadherin-4/R-Cadherin | |
| Polyclonal | |
| Unconjugated | |
| P39038 | |
| CDH4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cadherin-4/R-Cadherin His21-Ala731 Accession # P39038 | |
| 100 μg | |
| Primary | |
| Detects mouse Cadherin-4/R-Cadherin in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human Cadherin-4/R-Cadherin and recombinant mouse (rm) Cadherin-2 is observed and less than 1% cross-reactivity with rmE-Cadherin is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, Cadherin4, CDH4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1002 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction