missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse C5L2/GPR77 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC4729T-100UG
This item is not returnable.
View return policy
Description
C5L2/GPR77 Monoclonal specifically detects C5L2/GPR77 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| C5L2/GPR77 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8BW93 | |
| C5ar2 | |
| Chinese hamster ovary cell line transfected with mouse C5L2 Met1-Val344 Accession # Q8BW93 | |
| RUO | |
| 27202 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 468705 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77, GPR77 | |
| Rat | |
| 100 μg | |
| Primary | |
| This antibody stains mouse C5L2 transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction