missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse C4.4A/LYPD3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5567-SP
This item is not returnable.
View return policy
Description
C4.4A/LYPD3 Polyclonal specifically detects C4.4A/LYPD3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| C4.4A/LYPD3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, LYPD3, Matrigel-induced gene C4 protein, MIG-C4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27076 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q91YK8 | |
| LYPD3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse C4.4A Leu33-His287 Accession # Q91YK8 | |
| 25 μg | |
| Primary | |
| Detects mouse C4.4A in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human C4.4A is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction