missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse c-Rel Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2699
This item is not returnable.
View return policy
Description
c-Rel Monoclonal specifically detects c-Rel in Mouse samples. It is validated for Western Blot.
Specifications
| c-Rel | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P15307 | |
| REL | |
| E. coli-derived recombinant mouse c-Rel Met1-Ile588 Accession # P15307 | |
| 100 μg | |
| Primary | |
| Detects endogenous mouse c-Rel in Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 290512 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cRel, C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, REL, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5966 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction