missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BTNL2/Butyrophilin-like 2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5236
This item is not returnable.
View return policy
Description
BTNL2/Butyrophilin-like 2 Polyclonal specifically detects BTNL2/Butyrophilin-like 2 in Mouse samples. It is validated for Western Blot.
Specifications
| BTNL2/Butyrophilin-like 2 | |
| Polyclonal | |
| Unconjugated | |
| O70355 | |
| BTNL2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse BTNL2/Butyrophilin-like 2 aa 27-452 Accession # O70355 | |
| 100 μg | |
| Primary | |
| Detects mouse BTNL2/Butyrophilin-like 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) BTN, rmBTN2, and recombinant human BTN is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| BTL-II, Butyrophilin like 2, Butyrophilin-like 2, butyrophilin-like 2 (MHC class II associated) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 56244 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction