missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BOC Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2385
This item is not returnable.
View return policy
Description
BOC Polyclonal specifically detects BOC in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BOC | |
| Polyclonal | |
| Unconjugated | |
| AAH78631 | |
| BOC | |
| Mouse myeloma cell line NS0-derived recombinant mouse BOC Asp26-Lys508 (Gln ins) Val509-Asp846 Accession # AAH78631 | |
| 100 μg | |
| Primary | |
| Detects mouse BOC in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human BOC is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Boc homolog (mouse), brother of CDO, Protein BOC | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 91653 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction