missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse BMP-5 Recombinant Protein, PeproTech®

Product Code. p-200111260
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 30245123

Brand: Gibco™ 31542500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TGF-ß family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth, and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling, and maintenance of a variety of other tissues and organs. BMP-5 is expressed in the nervous system, lungs and liver. It is a known regulator for dendritic growth in sympathetic neurons. BMP-5 is a 454 amino acid precursor protein that is cleaved to release the biologically active C-terminal mature protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49003
For Use With (Application) Functional Assay
Formulation Protein with no preservative
Gene ID (Entrez) 12160
Molecular Weight (g/mol) 31.2 kDa
Name BMP5
Quantity 500 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias AU023399; BMP; BMP 5; Bmp5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein; bone morphogenetic protein 5; MGC34244; se; short ear
Gene Symbol Bmp5
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence AASKRKNQNR NKSNSHQDPS RMPSAGDYNT SEQKQACKKH ELYVSFRDLG WQDWIIAPEG YAAFYCDGEC SFPLNAHMNA TNHAIVQTLV HLMFPDHVPK PCCAPTKLNA ISVLYFDDSS NVILKKYRNM VVRSCGCH
Activity Determined by its ability to induce alkaline phosphatase production by ADTC-5 cells.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.