missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BMP-5 Antibody, R&D Systems™

Description
BMP-5 Monoclonal specifically detects BMP-5 in Mouse samples. It is validated for Neutralization.
Specifications
Specifications
| Antigen | BMP-5 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Clone | 781321 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Neutralization 5-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P49003 |
| Gene Alias | BMP5, BMP-5, bone morphogenetic protein 5, MGC34244 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?