missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BMP-5 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6176-SP
This item is not returnable.
View return policy
Description
BMP-5 Polyclonal specifically detects BMP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BMP-5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP5, BMP-5, bone morphogenetic protein 5, MGC34244 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 653 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P49003 | |
| BMP5 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse BMP-5 Ala315-His452 Accession # P49003 | |
| 25 μg | |
| Primary | |
| Detects mouse BMP-5 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human BMP-5 is observed. In direct ELISAs, approximately 100% cross-reactivity with recombinant human BMP-5 is observed, and approximately 4% cross-reactivity with recombinant mouse (rm) BMP-6 and rmBMP-7 is observed, and less than 1% cross-reactivity with rmBMP-8b is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction