missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BMP-15/GDF-9B Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB5917
This item is not returnable.
View return policy
Description
BMP-15/GDF-9B Monoclonal specifically detects BMP-15/GDF-9B in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| BMP-15/GDF-9B | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9Z0L4 | |
| BMP15 | |
| E. coli-derived recombinant human BMP-15/GDF-9B Gln268-Arg392 Accession # Q9Z0L4 | |
| 100 μg | |
| Primary | |
| Detects mouse BMP-15/GDF-9B in direct ELISAs. In this format, no cross-reactivity with recombinant human BMP1.1, 2, 3, 3b, 4, 5, 6, 7, 8, 9, 10, 15, recombinant mouse BMP-3, 3b, 4, 5, 6, 7, 8b, 9, or 10 is observed. | |
| Mouse | |
| Purified |
| Immunohistochemistry | |
| 582703 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP15, BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9210 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction