missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BMP-10 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB6038
This item is not returnable.
View return policy
Description
BMP-10 Monoclonal specifically detects BMP-10 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| BMP-10 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9R229 | |
| BMP10 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BMP-10 Asn314-Arg421 Accession # Q9R229 | |
| 100 μg | |
| Primary | |
| Detects mouse BMP-10 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) BMP-10 and less than 10% cross-reactivity with rhBMP-8a and recombinant mouse (rm) BMP-8b is observed. No cross-reactivity with rmBMP-3, -4, -5, -6, -7, -9, -15, rhBMP-1.1, -2, -3b, or the propeptide of rhBMP-10 is observed. | |
| Mouse | |
| Purified |
| Immunohistochemistry | |
| 654319 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP10, BMP-10, bone morphogenetic protein 10, MGC126783 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27302 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction