missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Betacellulin/BTC Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1025-SP
This item is not returnable.
View return policy
Description
Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot.
Specifications
| Betacellulin/BTC | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q05928 | |
| BTC | |
| E. coli-derived recombinant mouse Betacellulin/BTC Asp32-Gln118 Accession # Q05928 | |
| 25 μg | |
| Primary | |
| Detects Betacellulin/BTC in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Androgen R, rhBetacellulin, rhEGF, rhHB-EGF, rhHRG-alpha , rhHRG-beta 1, rhTGF-alpha , recombinant mouse (rm) Epigen, or rmEpiregulin is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 197415 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| betacellulin, BTC, probetacellulin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 685 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction