missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Betacellulin/BTC Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1025
This item is not returnable.
View return policy
Description
Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot.
Specifications
| Betacellulin/BTC | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q05928 | |
| BTC | |
| E. coli-derived recombinant mouse Betacellulin/BTC Asp32-Gln118 Accession # Q05928 | |
| 500 μg | |
| Primary | |
| Detects Betacellulin/BTC in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Androgen R, rhBetacellulin, rhEGF, rhHB-EGF, rhHRG-alpha , rhHRG-beta 1, rhTGF-alpha , recombinant mouse (rm) Epigen, or rmEpiregulin is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 197415 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| betacellulin, BTC, probetacellulin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 685 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction