missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse beta IG-H3 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2559
This item is not returnable.
View return policy
Description
Beta Ig-h3/TGFBI Monoclonal specifically detects Beta Ig-h3/TGFBI in Mouse samples. It is validated for Western Blot.
Specifications
| Beta Ig-h3/TGFBI | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P82198 | |
| TGFBI | |
| Mouse myeloma cell line NS0-derived recombinant mouse beta IG-H3 Gly24-His683 Accession # P82198 | |
| 100 μg | |
| Primary | |
| Detects mouse beta IG-H3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human beta IG-H3 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 320305 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta IGH3, Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, TGFBIP, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7045 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction