missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Mouse BAFF ELISA Kit
Shop All Proteintech Products
Click to view available options
Target Species:
Mouse
Unit Size:
96 tests
Description
- The assay recognizes mouse BAFF. An antibody specific for BAFF has been pre-coated onto the microwells.
- The BAFF protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for BAFF is added to detect the captured BAFF protein.
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm
- TNFSF13B, also named as BAFF, BLYS, TALL1, TNFSF20, ZTNF4 and CD257, is a cytokine that binds to TNFRSF13B/TACI and TNFRSF17/BCMA. TNFSF13B is localized in chromosome region 13q32-q34 and is implicated in the regulation of immune response to infections, autoimmune disease, and inflammation.
- This cytokine is expressed in B cell lineage cells, and acts as a potent B cell activator. It has been also shown to play an important role in the proliferation and differentiation of B cells.
- At the placental level, it is believed that TNFSF13B has an antiapoptotic effect.
- TNFSF13B is overexpressed in several autoimmune patients although the mechanism behind this feature is unclear.
Specifications
Specifications
| Accession Number | NP_001334238 |
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 23 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Mouse |
| Gene ID (Entrez) | 24099 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Mouse BAFF ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction