missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-H4 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2154
This item is not returnable.
View return policy
Description
B7-H4 Monoclonal specifically detects B7-H4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| B7-H4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q7TSP5 | |
| VTCN1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H4 Phe29-Pro258 Accession # Q7TSP5 | |
| 100 μg | |
| Primary | |
| Detects mouse B7-H4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H2, rmB7-H3, recombinant human (rh) B7-H3b, rhB7-H4, or rmPD-L2 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 297219 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79679 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction