missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-H2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB158
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Spezifikation
| B7-H2/ICOSLG | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q9JHJ8 | |
| Icosl | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H2 Glu47-Lys279 Accession # Q9JHJ8 | |
| 100 μg | |
| Primary | |
| Detects mouse B7-H2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-H2, rhB7-H3, rhB7-H3b, rhPD-L2, recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H3, rmB7-H4, rmPD-L2, or recombinant rat B7-2 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 599841 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23308 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur