missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-H2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF158-SP
This item is not returnable.
View return policy
Description
B7-H2/ICOSLG Polyclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| B7-H2/ICOSLG | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23308 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q9JHJ8 | |
| Icosl | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H2 Glu47-Lys279 Accession # Q9JHJ8 | |
| 25 μg | |
| Primary | |
| Detects mouse B7-H2 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction