missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-H1/PD-L1 APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB1019A-100
This item is not returnable.
View return policy
Description
PD-L1 Polyclonal specifically detects PD-L1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| PD-L1/B7-H1 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q9EP73 | |
| CD274 | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H1/PD-L1 Phe19-Thr238 Accession # Q9EP73 | |
| 100 Tests | |
| Primary | |
| Detects mouse B7-H1/PD-L1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human B7-H1 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29126 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction