missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-H1/PD-L1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB90781T-100UG
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal specifically detects PD-L1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| PD-L1/B7-H1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H1 Met1-His239 Accession # Q9EP73 | |
| RUO | |
| 29126 | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| 2096C | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9EP73 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects mouse B7-H1 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction