missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B220/CD45R Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1217
428.40 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
B220/CD45R Monoclonal antibody MAB1217 Specifically detects B220/CD45R in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Specifications
| B220/CD45R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PTPRC | |
| Abelson mouse leukemia virus-induced pre-B tumor cells | |
| 500 μg | |
| Primary | |
| Detects mouse B220/CD45R. | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| RA3-6B2 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation, Immunocytochemistry 8-25 ug/mL, CyTOF-reported | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5788 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction