missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse APLP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3685-SP
This item is not returnable.
View return policy
Description
APLP-2 Polyclonal specifically detects APLP-2 in Mouse samples. It is validated for Western Blot.
Specifications
| APLP-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP2, APLP-2, APPH, APPHCDEI box-binding protein, APPL2, CDEBP, WALPLP2, YWK-II | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 334 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AAH52396.2 | |
| APLP2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse APLP-2, Leu37-Ser624, Accession # AAH52396.2 | |
| 25 μg | |
| Primary | |
| Detects mouse APLP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) APLP-2 is observed and less than 1% cross-reactivity with rhAPLP-1 and recombinant mouse APLP-1 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction