Learn More
Mouse APLP-2 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF3685
 View more versions of this product
Product Code. 18023122
  • £451.00 / 100µg
Description
APLP-2 Polyclonal specifically detects APLP-2 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APLP-2
Polyclonal
Unconjugated
AAH52396.2
APLP2
Mouse myeloma cell line NS0-derived recombinant mouse APLP-2, Leu37-Ser624, Accession # AAH52396.2
100 μg
Primary
Detects mouse APLP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) APLP-2 is observed and less than 1% cross-reactivity with rhAPLP-1 and recombinant mouse APLP-1 is observed.
Mouse
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP2, APLP-2, APPH, APPHCDEI box-binding protein, APPL2, CDEBP, WALPLP2, YWK-II
Goat
Antigen Affinity-purified
RUO
334
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.