missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse APLP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
£147.00 - £410.00
Specifications
| Antigen | APLP-1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 25 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718077
![]() |
R&D Systems
AF3179-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18072652
![]() |
R&D Systems
AF3179 |
100 μg |
£410.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APLP-1 Polyclonal specifically detects APLP-1 in Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.Specifications
| APLP-1 | |
| Western Blot 0.1 ug/mL, Simple Western 25 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1 | |
| DPP4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q03157 | |
| 333 | |
| Mouse myeloma cell line NS0-derived recombinant mouse APLP-1 Gly37-Arg582 Accession # Q03157 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse APLP-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human APLP-1 is observed and less than 1% cross-reactivity with recombinant mouse APLP-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title