missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zp3r, Mouse, Clone: 7H12, Abnova™
Mouse monoclonal antibody raised against Zp3r.
Brand: Abnova MAB8386.100ug
This item is not returnable.
View return policy
Specifications
| Zp3r | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Zp3r | |
| Zp3r | |
| A mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient. | |
| RUO | |
| 22789 | |
| Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 7H12 | |
| Mouse monoclonal antibody raised against Zp3r. | |
| In PBS, pH 7.4 | |
| AV264399/SP56 | |
| Mouse | |
| 100 μg | |
| Primary | |
| These antibodies specifically recognize mouse sperm protein sp56. They will react with denatured, nonreduced sp56 but not with denatured, reduced sp56. These antibodies do not cross-react with human sperm. | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction