Learn More
ZNF274, Mouse, Clone: 4C12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZNF274.
Brand: Abnova H00010782-M01A.200uL
Description
This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals. [provided by RefSeq
Sequence: QKIDNPESQANSGALDTNQVLLHKIPPRKRLRKRDSQVKSMKHNSRVKIHQKSCERQKAKEGNGCRKTFSRSTKQITFIRIHKGSQVCRCSECGKIFRNPRYFSVHKKIHTSpecifications
| ZNF274 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC009763.2 | |
| ZNF274 | |
| ZNF274 (AAH09763.1,420 a.a. ∽ 530 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10782 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4C12 | |
| Mouse monoclonal antibody raised against a partial recombinant ZNF274. | |
| ZNF274 | |
| DKFZp686K08243/FLJ37843/HFB101/ZF2/ZKSCAN19 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.