Learn More
XPC, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant XPC.
Brand: Abnova H00007508-A01.50uL
Description
This gene encodes a component of the nucleotide excision repair (NER) pathway. There are multiple components involved in the NER pathway, including Xeroderma pigmentosum (XP) A-G and V, Cockayne syndrome (CS) A and B, and trichothiodystrophy (TTD) group A, etc. This component, XPC, plays an important role in the early steps of global genome NER, especially in damage recognition, open complex formation, and repair protein complex formation. Mutations in this gene or some other NER components result in Xeroderma pigmentosum, a rare autosomal recessive disorder characterized by increased sensitivity to sunlight with the development of carcinomas at an early age. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: EPVLGDVRESTAFSRSLLPVKPVEIEIETPEQAKTRERSEKIKLEFETYLRRAMKRFNKGVHEDTHKVHLLCLLANGFYRNNICSQPDLHAIGLSIIPARFTRVLPRDVDSpecifications
| XPC | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant XPC. | |
| XPC | |
| XP3/XPCC | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004628 | |
| XPC | |
| XPC (NP_004619, 141 a.a. ∽ 250 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 7508 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only