Learn More
WNT7B, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant WNT7B.
Brand: Abnova H00007477-A01.50uL
Description
This gene is a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein. [provided by RefSeq
Sequence: VRASRLRQPTFLRIKQLRSYQKPMETDLVYIEKSPNYCEEDAATGSVGTQGRLCNRTSPGADGCDTMCCGRGYNTHQYTKVWQCNCKFHWCCFVKCNTCSERTEVFTCKSpecifications
| WNT7B | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant WNT7B. | |
| WNT7B | |
| WNT7B | |
| WNT7B (NP_478679, 241 a.a. ∽ 349 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 7477 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_058238 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only