missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG1, Mouse, FITC, Clone: CT1, Abnova™
Mouse monoclonal antibody raised against native VSIG1.
Brand: Abnova MAB5856.500ug
This item is not returnable.
View return policy
Specifications
| VSIG1 | |
| Monoclonal | |
| FITC | |
| Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| VSIG1 | |
| VSIG1 | |
| Native purified from chicken thymocytes. | |
| RUO | |
| 414795 | |
| Chicken | |
| Liquid |
| Flow Cytometry, Immunoprecipitation | |
| CT1 | |
| Mouse monoclonal antibody raised against native VSIG1. | |
| In PBS (0.09% sodium azide) | |
| CHT1 | |
| Mouse | |
| 500 μg | |
| Primary | |
| ChT1 antigen (Mr 63/104 KDa) on chicken thymocytes. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction