missing translation for 'onlineSavingsMsg'
Learn More

VASN, Mouse, Clone: 4G7, Abnova™

Mouse monoclonal antibody raised against a partial recombinant VASN.

Brand:  Abnova H00114990-M05A.200uL

Product Code. 16161457

  • £303.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: NPFNCVCPLSWFGPWVRESHVTLASPEETRCHFPPKNAGRLLLELDYADFGC
Specifications

Specifications

VASN
Monoclonal
Unconjugated
ascites with no preservative
NM_138440
VASN
VASN (NP_612449,298 a.a. ∽ 349 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
114990
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Ascites
ELISA, Western Blot
4G7
Mouse monoclonal antibody raised against a partial recombinant VASN.
VASN
SLITL2
Mouse
200 μL
Primary
Human
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.